Learn More
Medchemexpress LLC 4-Morpholineethanesulfonic acid, potassium salt | 39946-25-3 | 99.9% | 10 G
SDP
Catalog No. 5000410600
Encompass_Preferred
MES potassium (2-Morpholinoethanesulphonic acid potassium) is an amphoteric ion buffer with a buffering capacity ranging from pH 5.5-7.0. As a Good's buffer, it is utilized in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies. This zwitterionic buffer provides effective buffering capacity around pH 6.15 and shows poor affinity with metal ions, making it suitable for studies involving them.
  • Amphoteric ion buffer
  • Buffer capacity ranging pH 5.5-7.0
  • Widely used in biochemistry and molecular biology experiments
  • Effective buffering capacity at around pH 6.15
  • Poor affinity with metal ions, making it suitable for studies involving metal ions
Catalog No. 50-004-10600 Supplier Medchemexpress LLC Supplier No. HYD0858E10G
Add to Cart
Add to Cart
MES potassium (2-Morpholinoethanesulphonic acid potassium) is an amphoteric ion buffer with a buffering capacity ranging from pH 5.5-7.0. As a Good's buffer, it is utilized in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies. This zwitterionic buffer provides effective buffering capacity around pH 6.15 and shows poor affinity with metal ions, making it suitable for studies involving them.
  • Amphoteric ion buffer
  • Buffer capacity ranging pH 5.5-7.0
  • Widely used in biochemistry and molecular biology experiments
  • Effective buffering capacity at around pH 6.15
  • Poor affinity with metal ions, making it suitable for studies involving metal ions