Learn More
Medchemexpress LLC 4-morpholineethanesulfonic acid, sodium salt (1:1) | 71119-23-8 | 98.0% | 10 G
SDP
Catalog No. 5000410158
Encompass_Preferred
MES sodium salt (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer with a buffering capacity ranging from pH 5.5-7.0. It is classified as a Good's buffer and is widely employed in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Amphoteric ion buffer with buffer capacity ranging from pH 5.5-7.0.
  • Good's buffer.
  • Widely used in biochemistry and molecular biology experiments.
  • Provides effective buffering capacity around pH 6.15.
  • pKa value changes with temperature, which should be considered for experiments at different temperatures.
  • Shows good buffering properties in biological experiments.
  • Exhibits poor affinity with metal ions (binding constants at 20 °C and 0.1 M: Mg2+, 0.8; Ca2+, 0.8; Mn2+, 1.7), making it suitable for studies involving metal ions.
Catalog No. 50-004-10158 Supplier Medchemexpress LLC Supplier No. HYD0858B10G
Add to Cart
Add to Cart
MES sodium salt (2-Morpholinoethanesulphonic acid) is an amphoteric ion buffer with a buffering capacity ranging from pH 5.5-7.0. It is classified as a Good's buffer and is widely employed in biochemistry and molecular biology experiments, including cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Amphoteric ion buffer with buffer capacity ranging from pH 5.5-7.0.
  • Good's buffer.
  • Widely used in biochemistry and molecular biology experiments.
  • Provides effective buffering capacity around pH 6.15.
  • pKa value changes with temperature, which should be considered for experiments at different temperatures.
  • Shows good buffering properties in biological experiments.
  • Exhibits poor affinity with metal ions (binding constants at 20 °C and 0.1 M: Mg2+, 0.8; Ca2+, 0.8; Mn2+, 1.7), making it suitable for studies involving metal ions.