Learn More
Medchemexpress LLC 4-Morpholineethanesulfonic acid, sodium salt (2:1) | 117961-21-4 | 98.0% | 25 G
SDP
Catalog No. 5000414211
Encompass_Preferred
MES (2-Morpholinoethanesulphonic acid) hemisodium is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5 to 7.0. It is a Good's buffer widely used in biochemistry and molecular biology experiments.
  • Amphoteric ion buffer.
  • Buffer capacity ranging pH 5.5-7.0.
  • Widely used in biochemistry and molecular biology experiments, such as cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Zwitterionic buffer, providing effective buffering capacity around pH 6.15.
  • pKa value changes with temperature, requiring consideration for different temperatures.
  • Shows good buffering properties in biological experiments.
  • Exhibits poor affinity with metal ions (binding constants at 20 °C and 0.1 M: Mg2+, 0.8; Ca2+, 0.8; Mn2+, 1.7), making it suitable for studies involving metal ions.
Catalog No. 50-004-14211 Supplier Medchemexpress LLC Supplier No. 25GHYD0858D
Add to Cart
Add to Cart
MES (2-Morpholinoethanesulphonic acid) hemisodium is an amphoteric ion buffer with a buffer capacity ranging from pH 5.5 to 7.0. It is a Good's buffer widely used in biochemistry and molecular biology experiments.
  • Amphoteric ion buffer.
  • Buffer capacity ranging pH 5.5-7.0.
  • Widely used in biochemistry and molecular biology experiments, such as cell culture, enzyme activity determination, electrophoresis, and protein studies.
  • Zwitterionic buffer, providing effective buffering capacity around pH 6.15.
  • pKa value changes with temperature, requiring consideration for different temperatures.
  • Shows good buffering properties in biological experiments.
  • Exhibits poor affinity with metal ions (binding constants at 20 °C and 0.1 M: Mg2+, 0.8; Ca2+, 0.8; Mn2+, 1.7), making it suitable for studies involving metal ions.