Learn More
Medchemexpress LLC 4-Morpholinepropanesulfonic acid, sodium salt (1:1) | 71119-22-7 | 99.8% | 50 G
SDP
Catalog No. 5000410167
Encompass_Preferred
MOPS sodium salt is a buffering agent utilized in biological applications. It effectively maintains the pH of mammalian cell culture media. Additionally, MOPS sodium salt has been shown to significantly enhance the thermal stability of BSA (Bovine Serum Albumin).
  • Buffering agent for biological applications.
  • Maintains the pH of mammalian cell culture media.
  • Significantly enhances the thermal stability of BSA.
Catalog No. 50-004-10167 Supplier Medchemexpress LLC Supplier No. HYD085950G
MOPS sodium salt is a buffering agent utilized in biological applications. It effectively maintains the pH of mammalian cell culture media. Additionally, MOPS sodium salt has been shown to significantly enhance the thermal stability of BSA (Bovine Serum Albumin).
  • Buffering agent for biological applications.
  • Maintains the pH of mammalian cell culture media.
  • Significantly enhances the thermal stability of BSA.