Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-morpholinophenylboronic acid

Catalog No. CDS005076 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005076 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005076 Supplier Sigma Aldrich Supplier No. CDS005076100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Ethyl valerate is also known as the green apple flavor
  • Microwave spectrum of ethyl valerate, observed by molecular beam fourier transform microwave spectroscopy, has been reported.ethyl valerate has wide applications in the areas of food, pharmaceuticals and cosmetics industries

Specifications

Molecular Formula C10H14BNO3
Linear Formula C10H14BNO3
MDL Number MFCD03095169
Quantity 500 mL
Molecular Weight (g/mol) 207.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.