Learn More
Cayman Chemical 4-MTA (hydrochloride), Purity: ≥98%, CAS: 94784-92-6, 10 mg
SDP
Catalog No. 502952120
Encompass_Preferred
A selective serotonergic agent which increases serotonin release, blocks its re-uptake (IC50 = 102 nM), and inhibits MAO A; intended for research and forensic applications
Catalog No. 50-295-2120 Supplier Cayman Chemical Supplier No. 1186210 MG
A selective serotonergic agent which increases serotonin release, blocks its re-uptake (IC50 = 102 nM), and inhibits MAO A; intended for research and forensic applications