Learn More
Research Products International Corp 4-Methylumbelliferyl-a-D-mannopyranoside, 100 Milligrams
SDP
Catalog No. 50213489
Encompass_Preferred
Fluorescent substrate for α-D-Mannosidase. 4-Methylumbelliferone: <50ppm.
Catalog No. 50-213-489 Supplier Research Products International Corp Supplier No. M666000.1
Fluorescent substrate for α-D-Mannosidase. 4-Methylumbelliferone: <50ppm.