Learn More
Research Products International Corp 4-Methylumbelliferyl-B-D-Xylopyranoside, 1 Gram
SDP
Catalog No. 50213486
Encompass_Preferred
Fluorescent substrate for β-D-Xylosidase.
4-Methylumbelliferone: <50ppm
Catalog No. 50-213-486 Supplier Research Products International Corp Supplier No. M659351.0
Fluorescent substrate for β-D-Xylosidase.
4-Methylumbelliferone: <50ppm