Learn More
Research Products International Corp 4-Methylumbelliferyl-a-L-Arabinopyranoside, 100 Milligrams
SDP
Catalog No. 50213465
Encompass_Preferred
Fluorescent substrate for α-L-Arabinosidase. 4-Methylumbelliferone < 50ppm.
Catalog No. 50-213-465 Supplier Research Products International Corp Supplier No. M641550.1
Fluorescent substrate for α-L-Arabinosidase. 4-Methylumbelliferone < 50ppm.