Learn More
Research Products International Corp 4-Methylumbelliferyl-2-acetamido-2-deoxy-B-D-galactopyranoside [4-Methylumbelliferyl-N-acetyl-B-D-galactosaminide], 100 Milligrams
SDP
Catalog No. 50213460
Encompass_Preferred
Fluorescent substrate for N-Acetyl-β-D-galactosaminidase.
Catalog No. 50-213-460 Supplier Research Products International Corp Supplier No. M639000.1
Fluorescent substrate for N-Acetyl-β-D-galactosaminidase.