Learn More
Research Products International Corp 4-Methylumbelliferyl-B-D-Galactopyranoside [4-Methylumbelliferyl-B-D-Galactoside], 1 Gram
SDP
Catalog No. 50213473
Encompass_Preferred
Fluorescent substrate for β-D-Galactosidase.
  • 4-Methylumbelliferone <50ppm
Catalog No. 50-213-473 Supplier Research Products International Corp Supplier No. M655001.0
Fluorescent substrate for β-D-Galactosidase.
  • 4-Methylumbelliferone <50ppm