Learn More
Research Products International Corp 4-Methylumbelliferyl-2-acetamido-2-deoxy-B-D-glucopyranoside [4-Methylumbelliferyl-N-acetyl-B-D-glucosaminide], 1 Gram
SDP
Catalog No. 50213462
Encompass_Preferred
Fluorescent substrate for N-Acetyl-B-D-glucosaminidase and B-N-Acetyl-D-hexosaminidase. Ultrasensitive. Checked by actual enzymatic assay.
Catalog No. 50-213-462 Supplier Research Products International Corp Supplier No. M641001.0
Fluorescent substrate for N-Acetyl-B-D-glucosaminidase and B-N-Acetyl-D-hexosaminidase. Ultrasensitive. Checked by actual enzymatic assay.