Learn More
Research Products International Corp 4-Methylumbelliferyl-B-D-Galactopyranoside-6-Sulfate, Sodium Salt, 25 Milligrams
SDP
Catalog No. 50213475
Encompass_Preferred
Fluorescent substrate for Morquio A Sulphatase.
  • 4-Methylumbelliferone: <100PPM
Catalog No. 50-213-475 Supplier Research Products International Corp Supplier No. M656000.025
Fluorescent substrate for Morquio A Sulphatase.
  • 4-Methylumbelliferone: <100PPM