Learn More
Research Products International Corp 4-Methylumbelliferyl-2-Acetamido-2-Deoxy-6-Sulfate-B-D-Glucopyranoside Sodium Salt, 25 Milligrams
SDP
Catalog No. 50213464
Encompass_Preferred
Fluorescent substrate for N-Acetyl-β-D-glucosaminidase (Hexosaminidase A). Allows more rapid detection of Tay-Sachs.
4-Methylumbelliferone < 50ppm
Nonsulfated Derivative: None Detected (NMR, TLC)
Checked by enzymatic assay
Catalog No. 50-213-464 Supplier Research Products International Corp Supplier No. M641500.025
Fluorescent substrate for N-Acetyl-β-D-glucosaminidase (Hexosaminidase A). Allows more rapid detection of Tay-Sachs.
4-Methylumbelliferone < 50ppm
Nonsulfated Derivative: None Detected (NMR, TLC)
Checked by enzymatic assay