Learn More
Research Products International Corp 4-Methylumbelliferyl-a-L-Fucopyranoside, 25 Milligrams
SDP
Catalog No. 50213470
Encompass_Preferred
Fluorescent substrate for α-L-Fucosidase.
4-Methylumbelliferone <50ppm.
Catalog No. 50-213-470 Supplier Research Products International Corp Supplier No. M652000.025
Fluorescent substrate for α-L-Fucosidase.
4-Methylumbelliferone <50ppm.