Learn More
eMolecules​ 192130-34-0 | 4-N-(2-Aminoethyl)-1-N-Boc-piperazine | Combi-Blocks | MFCD02683049 | 229.324 | C11H23N3O2 | 98.000 | CC(C)(C)OC(=O)N1CCN(CCN)CC1 | 25g | 232307048
Catalog No. NC2038842 Shop All eMolecules Inc. Products
Encompass_Preferred
4-N-(2-Aminoethyl)-1-N-Boc-piperazine | Combi-Blocks | 192130-34-0 | MFCD02683049 | 229.324 | C11H23N3O2 | 98.000 | CC(C)(C)OC(=O)N1CCN(CCN)CC1 | 25g | 232307048
Catalog No. NC2038842 Supplier eMolecules​ Supplier No. 232307048
4-N-(2-Aminoethyl)-1-N-Boc-piperazine | Combi-Blocks | 192130-34-0 | MFCD02683049 | 229.324 | C11H23N3O2 | 98.000 | CC(C)(C)OC(=O)N1CCN(CCN)CC1 | 25g | 232307048