Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(N-[3-chlorophenyl]-carbamoyloxy)-2-butynyltrimethylammonium chloride | 55-45-8 | MFCD00055068 | 99.9% | 317.2 g/mol | C14H18Cl2N2O2 | 1 ML
SDP

Catalog No. 5000240180
Encompass_Preferred

McN-A-343 is a selective muscarinic M1 receptor agonist supplied as a ready-to-use research reagent for studying muscarinic signaling in biochemical, cellular, and ex vivo assays.

  • Selective M1 muscarinic receptor agonist for receptor pharmacology studies.
  • Supplied as a 10 mM solution in a 1 mL vial for convenient dosing.
  • High purity (99.9%) suitable for biochemical and cellular assays.
  • Molecular weight 317.2 g/mol and formula C14H18Cl2N2O2 for reference.
  • Soluble in aqueous media at experimental concentrations.
  • Provided with documentation including purity data and safety information.

Catalog No. 50-002-40180 Supplier Medchemexpress LLC Supplier No. HY10764810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

McN-A-343 is a selective muscarinic M1 receptor agonist supplied as a ready-to-use research reagent for studying muscarinic signaling in biochemical, cellular, and ex vivo assays.

  • Selective M1 muscarinic receptor agonist for receptor pharmacology studies.
  • Supplied as a 10 mM solution in a 1 mL vial for convenient dosing.
  • High purity (99.9%) suitable for biochemical and cellular assays.
  • Molecular weight 317.2 g/mol and formula C14H18Cl2N2O2 for reference.
  • Soluble in aqueous media at experimental concentrations.
  • Provided with documentation including purity data and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.