Learn More
Medchemexpress LLC 4-(N-Boc-amino)piperidine | 73874-95-0 | MFCD00798171 | 96% | 200.28 g/mol | 50 G
SDP
Catalog No. 5000404596
Encompass_Preferred
4-(N-Boc-amino)piperidine is a chemical compound with a molecular weight of 200.28 g/mol and a typical purity of 96%. It is intended for testing and research purposes only, not for use as pharmaceuticals, food products, or household goods. It is classified as an irritant, causing skin and serious eye irritation, and may cause respiratory irritation.
  • Used for testing and research purposes only
  • Molecular formula: C10H20N2O2
  • Known to cause skin and serious eye irritation
  • May cause respiratory irritation
  • Should be stored refrigerated
Catalog No. 50-004-04596 Supplier Medchemexpress LLC Supplier No. HY4000250G
4-(N-Boc-amino)piperidine is a chemical compound with a molecular weight of 200.28 g/mol and a typical purity of 96%. It is intended for testing and research purposes only, not for use as pharmaceuticals, food products, or household goods. It is classified as an irritant, causing skin and serious eye irritation, and may cause respiratory irritation.
  • Used for testing and research purposes only
  • Molecular formula: C10H20N2O2
  • Known to cause skin and serious eye irritation
  • May cause respiratory irritation
  • Should be stored refrigerated