Learn More
Cayman Chemical 4-(N-Boc-amino)piperidine, Purity: ≥95%, CAS: 73874-95-0, 5 g
SDP
Catalog No. 502978971
Encompass_Preferred
An organic building block; has been used in the synthesis of antiviral CCR5 antagonists and antibacterial agents
Catalog No. 50-297-8971 Supplier Cayman Chemical Supplier No. 303985 G
An organic building block; has been used in the synthesis of antiviral CCR5 antagonists and antibacterial agents