Learn More
Medchemexpress LLC 4-n-butylresorcinol | 18979-61-8 | MFCD01684800 | 99.5% | 166.22 g/mol | C10H14O2 | 1 ML
SDP
Catalog No. 5000240711
Encompass_Preferred
4-Butylresorcinol is a phenolic small molecule used as a tyrosinase inhibitor in biochemical and dermatological research. It has a reported IC50 of 11.27 μM against tyrosinase and is offered in both solid form and ready-to-use solutions (for example, 10 mM in DMSO) for assay and formulation work.
  • High purity (99.5%) suitable for analytical and biochemical assays.
  • Inhibits tyrosinase with a reported IC50 of 11.27 μM.
  • Available as solid powder and as a 10 mM solution in DMSO for convenience.
  • Molecular formula C10H14O2 and molecular weight 166.22 g/mol.
  • Multiple pack sizes available to support small-scale and bulk studies.
Catalog No. 50-002-40711 Supplier Medchemexpress LLC Supplier No. HY10736910MM 1ML
4-Butylresorcinol is a phenolic small molecule used as a tyrosinase inhibitor in biochemical and dermatological research. It has a reported IC50 of 11.27 μM against tyrosinase and is offered in both solid form and ready-to-use solutions (for example, 10 mM in DMSO) for assay and formulation work.
  • High purity (99.5%) suitable for analytical and biochemical assays.
  • Inhibits tyrosinase with a reported IC50 of 11.27 μM.
  • Available as solid powder and as a 10 mM solution in DMSO for convenience.
  • Molecular formula C10H14O2 and molecular weight 166.22 g/mol.
  • Multiple pack sizes available to support small-scale and bulk studies.