Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-[N-Ethyl-N-(2-methylphenyl)sulfamoyl]acetanilide, 97%, Thermo Scientific™

Catalog No. AAH60904MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60904MD 250 mg
AAH6090403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60904MD Supplier Thermo Scientific Chemicals Supplier No. H60904MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333430-43-6
Molecular Formula C17H20N2O3S
Molecular Weight (g/mol) 332.42
MDL Number MFCD01213966
InChI Key NGMYZQIEURPXLI-UHFFFAOYSA-N
Synonym n-4-ethyl 2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-o-tolyl-sulfamoyl-phenyl-acetamide, 4'-n-ethyl-n-2-methylphenyl sulfamoyl acetanilide, n-4-ethyl-2-methylphenyl sulfamoyl phenyl acetamide, n-4-ethyl-2-methylanilino sulfonyl phenyl acetamide, n-4-ethyl 2-methylphenyl amino sulfonyl phenyl acetamide
PubChem CID 743585
IUPAC Name N-[4-[ethyl-(2-methylphenyl)sulfamoyl]phenyl]acetamide
SMILES CCN(C1=CC=CC=C1C)S(=O)(=O)C1=CC=C(NC(C)=O)C=C1

Specifications

Quantity 250 mg
Formula Weight 332.42
Percent Purity 97%
Chemical Name or Material 4'-[N-Ethyl-N-(2-methylphenyl)sulfamoyl]acetanilide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.