Learn More
Medchemexpress LLC 4-(N-iodoacetamide)benzophenone | 76809-63-7 | MFCD07437855 | 95.0% | 365.17 | C15H12INO2 | 100 MG
SDP
Catalog No. 5000284474
Encompass_Preferred
4-(N-Iodoacetamide)benzophenone is an iodoacetamide-functionalized benzophenone used as a heterobifunctional photocrosslinking and sulfhydryl-reactive reagent in biochemical research. It is provided as a solid for laboratory use with reported purity of 95.0% and a molecular weight of 365.17 g/mol. Intended for research use only.
  • Sulfhydryl-reactive iodoacetamide group for cysteine labeling.
  • Photoreactive benzophenone moiety for UV-induced crosslinking.
  • Suitable for biochemical assays and proteomics workflows.
  • Available in small solid quantities for laboratory experiments.
  • High reported purity appropriate for research applications.
Catalog No. 50-002-84474 Supplier Medchemexpress LLC Supplier No. HYW559951100MG
4-(N-Iodoacetamide)benzophenone is an iodoacetamide-functionalized benzophenone used as a heterobifunctional photocrosslinking and sulfhydryl-reactive reagent in biochemical research. It is provided as a solid for laboratory use with reported purity of 95.0% and a molecular weight of 365.17 g/mol. Intended for research use only.
  • Sulfhydryl-reactive iodoacetamide group for cysteine labeling.
  • Photoreactive benzophenone moiety for UV-induced crosslinking.
  • Suitable for biochemical assays and proteomics workflows.
  • Available in small solid quantities for laboratory experiments.
  • High reported purity appropriate for research applications.