Learn More
Medchemexpress LLC 4-(N-iodoacetamide)benzophenone | 76809-63-7 | MFCD07437855 | 95.0% | 365.17 g/mol | C15H12INO2 | 25 MG
SDP
Catalog No. 5000284351
Encompass_Preferred
4-(N-Iodoacetamide)benzophenone is a sulfhydryl-reactive heterobifunctional reagent used in biochemical research for covalent labeling and photocrosslinking of proteins. It pairs an iodoacetamide electrophile that selectively alkylates cysteine thiols with a benzophenone photoreactive group that forms covalent bonds upon UV activation. The material is supplied as a solid in small research pack sizes and is accompanied by supporting documentation.
Catalog No. 50-002-84351 Supplier Medchemexpress LLC Supplier No. HYW55995125MG
4-(N-Iodoacetamide)benzophenone is a sulfhydryl-reactive heterobifunctional reagent used in biochemical research for covalent labeling and photocrosslinking of proteins. It pairs an iodoacetamide electrophile that selectively alkylates cysteine thiols with a benzophenone photoreactive group that forms covalent bonds upon UV activation. The material is supplied as a solid in small research pack sizes and is accompanied by supporting documentation.