Learn More
Sigma Aldrich Fine Chemicals Biosciences 4-N-Maleimidomethylcyclohe25MG
Catalog No. 502897577
Encompass
4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester (SMCC) is a heterobifunctional cross-linking reagent incorporating an extended spacer with amine and sulfhydryl reactivity. It is typically coupled initially to molecules containing primary amine by amide bond buffered at pH 7.5 (6.5-8.5). The second coupling is specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). SMCC contains nine atom linker. An extended aliphatic spacer stabilizes the maleimide prior to coupling compared to aromatic spacers.
Catalog No. 50-289-7577 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M552525MG
Add to Cart
Add to Cart
4-(N-Maleimidomethyl)cyclohexanecarboxylic acid N-hydroxysuccinimide ester (SMCC) is a heterobifunctional cross-linking reagent incorporating an extended spacer with amine and sulfhydryl reactivity. It is typically coupled initially to molecules containing primary amine by amide bond buffered at pH 7.5 (6.5-8.5). The second coupling is specific for molecules containing free sulfhydryl by thioether linkage buffered at pH 6.8 (6.5-7.0). SMCC contains nine atom linker. An extended aliphatic spacer stabilizes the maleimide prior to coupling compared to aromatic spacers.