Learn More
Strem, An Ascensus Company CAS# 932710-63-9. 1g. [4-(N,N-Dimethylamino)phenyl]di-t-butylphosphine, min. 95% amphos. MFCD09265102
SDP
Catalog No. NC0782181
Encompass
CAS# 932710-63-9. 1g. [4-(N,N-Dimethylamino)phenyl]di-t-butylphosphine, min. 95% amphos. MFCD09265102. Molecular Weight: 265.37. Molecular Formula: C16H28NP. Color/form: white to light-brown xtl. Strem# 15-1248. http://www.strem.com/catalog/v/15-1248/
Catalog No. NC0782181 Supplier Strem, An Ascensus Company Supplier No. 1512481G
CAS# 932710-63-9. 1g. [4-(N,N-Dimethylamino)phenyl]di-t-butylphosphine, min. 95% amphos. MFCD09265102. Molecular Weight: 265.37. Molecular Formula: C16H28NP. Color/form: white to light-brown xtl. Strem# 15-1248. http://www.strem.com/catalog/v/15-1248/