Learn More
Medchemexpress LLC 4-(N-Nitrosomethylamino)-1-(3-pyridyl)-1-butanone-d4 | 764661-24-7 | 99.7% | 1 MG
SDP
Catalog No. 5000342106
Encompass_Preferred
NNK-d4 is the deuterium labeled NNK. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-42106 Supplier Medchemexpress LLC Supplier No. HY126477S1MG
NNK-d4 is the deuterium labeled NNK. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS