Learn More
Medchemexpress LLC 4-(N-Nitrosomethylamino)-1-(3-pyridyl)-1-butanone-d4 | 764661-24-7 | 99.7% | 10 MG
SDP
Catalog No. 5000339156
Encompass_Preferred
NNK-d4 is a deuterium labeled compound, used as a tracer and internal standard for quantitative analysis by methods like NMR, GC-MS, or LC-MS. Its deuterium substitution can affect pharmacokinetic and metabolic profiles, making it valuable in drug development and research.
  • Used as a tracer and internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution may influence pharmacokinetic and metabolic profiles
  • White to off-white solid appearance
  • Purity of 99.65%
Catalog No. 50-003-39156 Supplier Medchemexpress LLC Supplier No. HY126477S10MG
NNK-d4 is a deuterium labeled compound, used as a tracer and internal standard for quantitative analysis by methods like NMR, GC-MS, or LC-MS. Its deuterium substitution can affect pharmacokinetic and metabolic profiles, making it valuable in drug development and research.
  • Used as a tracer and internal standard
  • Suitable for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium substitution may influence pharmacokinetic and metabolic profiles
  • White to off-white solid appearance
  • Purity of 99.65%