Learn More
Medchemexpress LLC 4-(N-nitrosomethylamino)-1-(3-pyridyl)-1-butanone-d4 | 764661-24-7 | 99.7% | 211.25 g/mol | C10H9D4N3O2 | 5 MG
SDP
Catalog No. 5000264544
Encompass_Preferred
NNK-d4 is the deuterium-labeled analogue of the tobacco-specific nitrosamine NNK, provided for research use as an isotopic tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is intended for laboratory research only and not for human use.
  • High purity (99.65%).
  • Isotopically labeled to improve quantitation and traceability in mass spectrometry.
  • Soluble in DMSO at ≥ 62.5 mg/mL for easy sample preparation.
  • Suitable as a tracer or internal standard for NMR, GC-MS, and LC-MS workflows.
  • Provided with supporting documentation including COA and SDS.
Catalog No. 50-002-64544 Supplier Medchemexpress LLC Supplier No. HY126477S5MG
NNK-d4 is the deuterium-labeled analogue of the tobacco-specific nitrosamine NNK, provided for research use as an isotopic tracer or internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It is intended for laboratory research only and not for human use.
  • High purity (99.65%).
  • Isotopically labeled to improve quantitation and traceability in mass spectrometry.
  • Soluble in DMSO at ≥ 62.5 mg/mL for easy sample preparation.
  • Suitable as a tracer or internal standard for NMR, GC-MS, and LC-MS workflows.
  • Provided with supporting documentation including COA and SDS.