Learn More
Cayman Chemical 4- n-nonyl BenzenboronIc Ac 1g
SDP
Catalog No. 502952727
Encompass_Preferred
An intermediary prodrug of fluorouracil the pyrimidine analog that inhibits thymidylate synthase selectively in tumor cells interfering with DNA synthesis one of two intermediary metabolites formed during metabolic conversion of the chemotherapeutic prodrug capecitabine to fluorouracil
Catalog No. 50-295-2727 Supplier Cayman Chemical Supplier No. 131401 G
An intermediary prodrug of fluorouracil the pyrimidine analog that inhibits thymidylate synthase selectively in tumor cells interfering with DNA synthesis one of two intermediary metabolites formed during metabolic conversion of the chemotherapeutic prodrug capecitabine to fluorouracil