Learn More
Cayman Chemical 4- n-nonyl Benzeneboronic Ac
SDP
Catalog No. 502952762
Encompass_Preferred
An analog of JWH 018 a selective agonist of the CB1 receptor intended for research and forensic applications
Catalog No. 50-295-2762 Supplier Cayman Chemical Supplier No. 13140500 MG
An analog of JWH 018 a selective agonist of the CB1 receptor intended for research and forensic applications