Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Octyl-4H-dithieno[3,2-b:2',3'-d]pyrrole 98.0+%, TCI America™
SDP

Catalog No. O04311G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04311G 1 g
O0431200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04311G Supplier TCI America Supplier No. O04311G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141029-75-6
Molecular Formula C16H21NS2
Molecular Weight (g/mol) 291.47
MDL Number MFCD26793506
InChI Key GGCJLBJHTUIOES-UHFFFAOYSA-N
PubChem CID 11033644
IUPAC Name 7-octyl-3,11-dithia-7-azatricyclo[6.3.0.0²,⁶]undeca-1(8),2(6),4,9-tetraene
SMILES CCCCCCCCN1C2=C(SC=C2)C2=C1C=CS2

Specifications

Melting Point 35°C
Color White-Yellow
Quantity 1 g
Formula Weight 291.47
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-n-Octyl-4H-dithieno[3,2-b:2′,3′-d]pyrrole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.