Learn More
Medchemexpress LLC 4-n-octyl-anisole-d17 | 1219794-56-5 | 99.2% | 237.46 | C15H7D17O | 10 MG
SDP
Supplier:  Medchemexpress LLC HY151186S10MG
Encompass_Preferred
4-n-Octyl-anisole-d17 is the deuterium-labeled analogue of 4-n-octyl-anisole intended for research use as a tracer and internal standard in quantitative analytical methods. It is used in pharmacokinetic and metabolic studies and is compatible with NMR, GC-MS, and LC-MS workflows.
  • Deuterium labeling (d17) provides a clear mass shift for tracer and internal standard applications.
  • Compatible with NMR, GC-MS, and LC-MS for quantitative analysis.
  • Stable isotope labeling supports pharmacokinetic and metabolic profiling.
  • Chemical formula C15H7D17O and molecular weight 237.46 aid molecular identification.
  • High reported purity (99.2%) suitable for analytical standards.
  • Available in small research package sizes (1 mg, 5 mg, 10 mg).
Catalog No. 50-002-87292