Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Octylstyrene (stabilized with TBC) 95.0+%, TCI America™
SDP

Catalog No. O02735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02735G Supplier TCI America Supplier No. O02735G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 46745-66-8
Molecular Formula C16H24
Molecular Weight (g/mol) 216.368
MDL Number MFCD00078353
InChI Key HLRQDIVVLOCZPH-UHFFFAOYSA-N
PubChem CID 17918559
IUPAC Name 1-ethenyl-4-octylbenzene
SMILES CCCCCCCCC1=CC=C(C=C1)C=C

Specifications

Color Yellow
Boiling Point 114°C
Quantity 5 g
Formula Weight 216.37
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-n-Octylstyrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.