Learn More
Medchemexpress LLC 4-naphthalen-1-yl-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinoline-8-sulfonamide | 353483-92-8 | MFCD01080566 | 99.8% | 376.47 | C22H20N2O2S | 100 MG
SDP
Catalog No. 5000240277
Encompass_Preferred
TQS is a small-molecule positive allosteric modulator of the α7 nicotinic acetylcholine receptor provided for research use. It is an off-white to light-yellow solid with characterized physicochemical properties, solubility profiles, and recommended storage conditions for both powder and stock solutions.
  • Positive allosteric modulator of α7 nicotinic acetylcholine receptor.
  • Suitable for in vitro and in vivo neuroinflammatory pain research.
  • High reported purity (99.84%).
  • Chemical formula C22H20N2O2S; molecular weight 376.47.
  • Highly soluble in DMSO (125 mg/mL) for stock solutions.
  • Stable when stored as powder at -20°C or lower; stock solutions stable at -80°C for months.
Catalog No. 50-002-40277 Supplier Medchemexpress LLC Supplier No. HY107682100MG
TQS is a small-molecule positive allosteric modulator of the α7 nicotinic acetylcholine receptor provided for research use. It is an off-white to light-yellow solid with characterized physicochemical properties, solubility profiles, and recommended storage conditions for both powder and stock solutions.
  • Positive allosteric modulator of α7 nicotinic acetylcholine receptor.
  • Suitable for in vitro and in vivo neuroinflammatory pain research.
  • High reported purity (99.84%).
  • Chemical formula C22H20N2O2S; molecular weight 376.47.
  • Highly soluble in DMSO (125 mg/mL) for stock solutions.
  • Stable when stored as powder at -20°C or lower; stock solutions stable at -80°C for months.