Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-naphthalen-1-yl-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinoline-8-sulfonamide | 353483-92-8 | MFCD01080566 | 99.8% | 376.47 | C22H20N2O2S | 50 MG
SDP

Catalog No. 5000240482
Encompass_Preferred

TQS is a selective positive allosteric modulator of the α7 nicotinic acetylcholine receptor (nAChR) used in preclinical research, including studies of neuroinflammatory pain. It is provided as a high-purity research chemical and is available as a powder or as a DMSO solution for experimental use.

  • Selective α7 positive allosteric modulator for receptor modulation studies.
  • Suitable for in vitro and in vivo neuroinflammatory pain research.
  • High purity (~99.8%).
  • Available as powder and as a 10 mM solution in DMSO.
  • Recommended storage: powder -20 °C long term or 4 °C short term; in solvent -80 °C.
  • Molecular formula C22H20N2O2S and molecular weight 376.47 g·mol-1.

Catalog No. 50-002-40482 Supplier Medchemexpress LLC Supplier No. HY10768250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TQS is a selective positive allosteric modulator of the α7 nicotinic acetylcholine receptor (nAChR) used in preclinical research, including studies of neuroinflammatory pain. It is provided as a high-purity research chemical and is available as a powder or as a DMSO solution for experimental use.

  • Selective α7 positive allosteric modulator for receptor modulation studies.
  • Suitable for in vitro and in vivo neuroinflammatory pain research.
  • High purity (~99.8%).
  • Available as powder and as a 10 mM solution in DMSO.
  • Recommended storage: powder -20 °C long term or 4 °C short term; in solvent -80 °C.
  • Molecular formula C22H20N2O2S and molecular weight 376.47 g·mol-1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.