Learn More
Medchemexpress LLC 4-naphthalen-1-yl-3a,4,5,9b-tetrahydro-3H-cyclopentan[c]quinoline-8-sulfonamide | 353483-92-8 | MFCD01080566 | >98.0% | 376.47 g/mol | C22H20N2O2S | 10 MG
SDP
Catalog No. 5000240019
Encompass_Preferred
TQS is a type II positive allosteric modulator of the α7 nicotinic acetylcholine receptor (α7 nAChR) used in preclinical research to potentiate agonist-evoked responses and reduce receptor desensitization. It is employed in studies of neuroinflammation and pain; chemically it is C22H20N2O2S with a molecular weight of 376.47 g/mol.
  • Enhances α7 receptor currents and reduces desensitization.
  • Type II positive allosteric modulation with pEC50 ≈ 5.5.
  • High reported purity (≥98% by HPLC).
  • Soluble in DMSO for solution preparations.
  • Supplied as solid or as DMSO solution for laboratory use.
Catalog No. 50-002-40019 Supplier Medchemexpress LLC Supplier No. HY10768210MG
TQS is a type II positive allosteric modulator of the α7 nicotinic acetylcholine receptor (α7 nAChR) used in preclinical research to potentiate agonist-evoked responses and reduce receptor desensitization. It is employed in studies of neuroinflammation and pain; chemically it is C22H20N2O2S with a molecular weight of 376.47 g/mol.
  • Enhances α7 receptor currents and reduces desensitization.
  • Type II positive allosteric modulation with pEC50 ≈ 5.5.
  • High reported purity (≥98% by HPLC).
  • Soluble in DMSO for solution preparations.
  • Supplied as solid or as DMSO solution for laboratory use.