Learn More
Medchemexpress LLC (4-nh2)-exatecan | 2495742-21-5 | 99.9% | C23H21N3O4 | 10MG
SDP
Catalog No. 5000261639
Encompass_Preferred
(4-NH2)-Exatecan is a 4-amino derivative of the topoisomerase inhibitor exatecan supplied as a high-purity research reagent. It is intended for use as a cytotoxic payload or intermediate in the synthesis of antibody-drug conjugates (ADCs), and is provided with storage guidance for solid and solution states.
  • Suitable as an antibody-drug conjugate (ADC) payload.
  • Amino group at the 4-position enables linker conjugation.
  • High purity appropriate for research applications.
  • Available in small to mid-scale pack sizes for medicinal chemistry.
  • Solid storage stability at 4°C when sealed and protected from moisture and light.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-61639 Supplier Medchemexpress LLC Supplier No. HY14539710MG
(4-NH2)-Exatecan is a 4-amino derivative of the topoisomerase inhibitor exatecan supplied as a high-purity research reagent. It is intended for use as a cytotoxic payload or intermediate in the synthesis of antibody-drug conjugates (ADCs), and is provided with storage guidance for solid and solution states.
  • Suitable as an antibody-drug conjugate (ADC) payload.
  • Amino group at the 4-position enables linker conjugation.
  • High purity appropriate for research applications.
  • Available in small to mid-scale pack sizes for medicinal chemistry.
  • Solid storage stability at 4°C when sealed and protected from moisture and light.
  • For research use only; not for human or clinical use.