Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-1H-pyrazole, 97%

Catalog No. ac430160050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC430160050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC430160050 Supplier Thermo Scientific Chemicals Supplier No. 430160050
Only null left
Add to Cart

CAS: 2075-46-9 | C3H3N3O2 | 113.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2075-46-9
Molecular Formula C3H3N3O2
Molecular Weight (g/mol) 113.08
MDL Number MFCD00159626
InChI Key XORHNJQEWQGXCN-UHFFFAOYSA-N
Synonym 4-nitropyrazole, 1h-pyrazole, 4-nitro, pyrazole, 4-nitro, 4-nitropyrrazole, 4-nitropyrazol, 4-nitro-pyrazole, 4-no2-pyrazole, 4-nitro-1h-pyrazol, 1h-pyrazole-4-nitro, pubchem14984
PubChem CID 16376
IUPAC Name 4-nitro-1H-pyrazole
SMILES [O-][N+](=O)C1=CNN=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 161.0°C to 162.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 113.08
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-1H-pyrazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.