Learn More
Medchemexpress LLC 4-nitro-2,1,3-benzoxadiazol-7-amine | 10199-91-4 | 96.7% | 25 MG
SDP
Catalog No. 5000231867
Encompass_Preferred
NBD-amine is a fluorogenic amine used as a fluorescent probe and labeling reagent. It fluoresces weakly in aqueous solution and strongly in organic solvents, lipid membranes, and other hydrophobic environments, making it suitable for sensing, labeling, and studies of membrane-associated processes.
  • Fluorogenic amine that fluoresces weakly in water and strongly in hydrophobic environments.
  • Useful as a fluorescent probe and labeling reagent for synthesis and detection.
  • Suitable for studying membrane interactions and lipid environments.
  • Provided as a solid with purity 96.7% and molecular weight 180.12 g/mol.
  • Documentation available: data sheet, safety data sheet (SDS), and certificate of analysis (COA).
Catalog No. 50-002-31867 Supplier Medchemexpress LLC Supplier No. HY11979125MG
NBD-amine is a fluorogenic amine used as a fluorescent probe and labeling reagent. It fluoresces weakly in aqueous solution and strongly in organic solvents, lipid membranes, and other hydrophobic environments, making it suitable for sensing, labeling, and studies of membrane-associated processes.
  • Fluorogenic amine that fluoresces weakly in water and strongly in hydrophobic environments.
  • Useful as a fluorescent probe and labeling reagent for synthesis and detection.
  • Suitable for studying membrane interactions and lipid environments.
  • Provided as a solid with purity 96.7% and molecular weight 180.12 g/mol.
  • Documentation available: data sheet, safety data sheet (SDS), and certificate of analysis (COA).