Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-2-(trifluoromethyl)aniline, 98%

Catalog No. AAL0097714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0097714 25 g
AAL0097706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0097714 Supplier Thermo Scientific Chemicals Supplier No. L0097714
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 121-01-7 | C7H5F3N2O2 | 206.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-01-7
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.124
MDL Number MFCD00007365
InChI Key HOTZLWVITTVZGY-UHFFFAOYSA-N
Synonym 2-amino-5-nitrobenzotrifluoride, 4-nitro-2-trifluoromethyl aniline, 2-amino-5-nitro-trifluorobenzene, 4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine, 4-nitro-2-trifluoromethyl benzenamine, 4-nitro-a,a,a-trifluoro-o-toluidine, benzenamine, 4-nitro-2-trifluoromethyl, 5-nitro-2-aminobenzotrifluoride, 2-trifluoromethyl-4-nitroaniline, a,a,a-trifluoro-4-nitro-o-toluidine
PubChem CID 67128
IUPAC Name 4-nitro-2-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N

Specifications

Melting Point 90°C to 93°C
Quantity 25 g
UN Number UN2811
Beilstein 2121347
Formula Weight 206.13
Percent Purity 98%
Chemical Name or Material 4-Nitro-2-(trifluoromethyl)aniline
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 204-443-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.