Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride, 97%

Catalog No. AAL1972106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1972106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1972106 Supplier Thermo Scientific Chemicals Supplier No. L1972106

CAS: 39234-83-8 | C7H3ClF3NO4S | 289.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39234-83-8
Molecular Formula C7H3ClF3NO4S
Molecular Weight (g/mol) 289.61
MDL Number MFCD03094387
InChI Key LJQUNSRHHHYXEW-UHFFFAOYSA-N
Synonym 4-nitro-3-trifluoromethyl benzenesulfonyl chloride, 4-nitro-3-trifluoromethyl benzenesulphonyl chloride, 4-nitro-3-trifluoromethyl benzenesulfonylchloride, 4-nitro-3-trifluoromethyl benzene-1-sulfonyl chloride, pubchem11722, acmc-1ail1, 5-chlorosulphonyl-2-nitrobenzotrifluoride, 4-nitro-3-trifluoromethyl phenylsulfonyl chloride, 4-chlorosulphonyl-2-trifluoromethyl nitrobenzene, 4-nitro-3-trifluoromethyl-benzenesulfonyl chloride
PubChem CID 2778595
IUPAC Name 4-nitro-3-(trifluoromethyl)benzenesulfonyl chloride
SMILES [O-][N+](=O)C1=C(C=C(C=C1)S(Cl)(=O)=O)C(F)(F)F

Specifications

Melting Point 63°C to 65°C
Quantity 5 g
UN Number UN3261
Beilstein 2880842
Sensitivity Moisture sensitive
Formula Weight 289.62
Percent Purity 97%
Chemical Name or Material 4-Nitro-3-(trifluoromethyl)benzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.