Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-nitro-DL-phenylalanine | 2922-40-9 | MFCD00007384 | 99.6% | 210.19 | C9H10N2O4 | 25 G
SDP

Catalog No. 5000303517
Encompass_Preferred

H-DL-Phe(4-NO2)-OH (4-nitro-DL-phenylalanine) is a nitro-substituted DL-phenylalanine derivative supplied as a high-purity solid for research use. It is used as a building block and reagent in synthetic chemistry and analytical applications.

  • Nitro-substituted DL-phenylalanine derivative for synthetic applications.
  • High purity (99.6%) suitable for analytical and synthesis work.
  • Molecular formula C9H10N2O4 and molecular weight 210.19.
  • Available in lab-scale pack sizes such as 25 g and 100 g.
  • Provided with supporting documentation including COA, SDS, HNMR, and LCMS for quality assurance.

Catalog No. 50-003-03517 Supplier Medchemexpress LLC Supplier No. 25GHYW014405
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-DL-Phe(4-NO2)-OH (4-nitro-DL-phenylalanine) is a nitro-substituted DL-phenylalanine derivative supplied as a high-purity solid for research use. It is used as a building block and reagent in synthetic chemistry and analytical applications.

  • Nitro-substituted DL-phenylalanine derivative for synthetic applications.
  • High purity (99.6%) suitable for analytical and synthesis work.
  • Molecular formula C9H10N2O4 and molecular weight 210.19.
  • Available in lab-scale pack sizes such as 25 g and 100 g.
  • Provided with supporting documentation including COA, SDS, HNMR, and LCMS for quality assurance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.