Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-DL-phenylalanine hydrate, 99%, Thermo Scientific™

Catalog No. AC225840050
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC225840050 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC225840050 Supplier Thermo Scientific Chemicals Supplier No. 225840050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2922-40-9
CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H10N2O4
Linear Formula O2NC6H4CH2CH(NH2)CO2H·xH2O
MDL Number MFCD00007384
Quantity 5g
Beilstein 14, 506
Synonym 4-nitro-dl-phenylalanine, 2-amino-3-4-nitrophenyl propanoic acid, 4-nitrophenylalanine, p-nitro-dl-phenylalanine, p-nitrophenylalanine, h-dl-phe 4-no2-oh, dl-p-nitrophenylalanine, h-p-nitro-dl-phe-oh, dl-phenylalanine, 4-nitro, 2-amino-3-4-nitrophenyl propionic acid
InChI Key GTVVZTAFGPQSPC-UHFFFAOYNA-N
SMILES NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O
IUPAC Name 2-amino-3-(4-nitrophenyl)propanoic acid
Molecular Weight (g/mol) 210.19
PubChem CID 65089
ChEBI CHEBI:48496
Formula Weight 210.19
Percent Purity ≥98.5%
Chemical Name or Material 4-Nitro-DL-phenylalanine hydrate, 99%
Show More Show Less
EINECSNumber 220-868-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.