Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-DL-phenylalanine Hydrate 98.0+%, TCI America™
SDP

Catalog No. N09525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09525G 5 g
N095225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09525G Supplier TCI America Supplier No. N09525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 207569-25-3
Molecular Formula C9H12N2O5
Molecular Weight (g/mol) 228.204
MDL Number MFCD00007384
InChI Key OLZDHJRNUIXKLU-UHFFFAOYSA-N
Synonym 2-Amino-3-(4-nitrophenyl)propionic Acid, H-DL-Phe(4-NO2)-OH
PubChem CID 18468090
IUPAC Name 2-amino-3-(4-nitrophenyl)propanoic acid;hydrate
SMILES C1=CC(=CC=C1CC(C(=O)O)N)[N+](=O)[O-].O

Specifications

Melting Point 230°C
Quantity 5 g
Formula Weight 210.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitro-DL-phenylalanine Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.