Learn More
Medchemexpress LLC 4-nitroacetophenone | 100-19-6 | MFCD00007355 | 99.7% | 165.15 g/mol | C8H7NO3 | 1kg
SDP
Catalog No. 5000269009
Encompass_Preferred
4 -Nitroacetophenone is a nitroaromatic compound 4 -Nitroacetophenone is an important organic synthetic intermediate which can be used for the synthesis of other active compounds such as Chloramphenicol (HY-B0239)[1][2][3]
Catalog No. 50-002-69009 Supplier Medchemexpress LLC Supplier No. HYY01851KG
Add to Cart
Add to Cart
4 -Nitroacetophenone is a nitroaromatic compound 4 -Nitroacetophenone is an important organic synthetic intermediate which can be used for the synthesis of other active compounds such as Chloramphenicol (HY-B0239)[1][2][3]