Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitroaniline, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020377 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0377 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0377 Supplier MilliporeSigma Supelco Supplier No. 31569250MG
Add to Cart
Add to Cart

4-Nitroaniline is an aromatic amine, used as an intermediate in the production of p-phenylenediamine, azo dyes, antioxidants, fuel additives, corrosion inhibitors and pesticides.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 100-01-6
Flash Point 213.0°C - Closed Cup
Physical Form Neat
Linear Formula O2 NC6H4 NH2
MDL Number MFCD00007858
Melting Point 146°C to 149°C
Boiling Point 260°C (100 mmHg)
Type Analytical Standard
Quantity 250 mg
Formula Weight 138.12
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.