Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitroaniline, ≥99.0% (HPLC), Solstice

Catalog No. 60047440 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-440 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-440 Supplier Solstice Supplier No. 72680500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Matrix substance for MALDI-MS,≥99.0% (HPLC)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-01-6
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007858
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
PubChem CID 7475
ChEBI CHEBI:17064
IUPAC Name 4-nitroaniline
SMILES NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 146°C to 149°C
Color Yellow
Density 1.44 g/cm3
pH 7
Boiling Point 260°C
Packaging Glass bottle
Linear Formula O2NC6H4NH2
Quantity 500 g
UN Number UN1661
Beilstein 508690
Formula Weight 138.12g/mol
Percent Purity ≥99.0% (HPLC)
Physical Form Crystals
Chemical Name or Material 4-Nitroaniline
Show More Show Less
Hazard Statement P261-P273-P280-P301 + P310-P311
DOTInformation 6.1 / PGII
RTECSNumber 202-810-1
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.