Learn More
Medchemexpress LLC 4-Nitrobenzaldehyde | 555-16-8 | 151.12 | 1 ML
SDP
Catalog No. 5000445966
Encompass_Preferred
4-Nitrobenzaldehyde is a widely used industrial chemical intermediate and also the main photodegradation product of the broad-spectrum antibiotic Chloramphenicol. It has genotoxic and mutagenic effects and poses a certain threat to human health and ecosystems. Research indicates that transgenic rice with the EINFS1 gene can convert 4-Nitrobenzaldehyde into 4-Aminobenzaldehyde with lower toxicity, showing application potential in the phytoremediation of 4-Nitrobenzaldehyde-contaminated environments.
  • Widely used industrial chemical intermediate
  • Main photodegradation product of the antibiotic Chloramphenicol
  • Has genotoxic and mutagenic effects
  • Poses a threat to human health and ecosystems
  • Can be converted to a less toxic compound by specific transgenic rice
  • Potential for use in phytoremediation of contaminated environments
Catalog No. 50-004-45966 Supplier Medchemexpress LLC Supplier No. HYW00757710MM/1ML
4-Nitrobenzaldehyde is a widely used industrial chemical intermediate and also the main photodegradation product of the broad-spectrum antibiotic Chloramphenicol. It has genotoxic and mutagenic effects and poses a certain threat to human health and ecosystems. Research indicates that transgenic rice with the EINFS1 gene can convert 4-Nitrobenzaldehyde into 4-Aminobenzaldehyde with lower toxicity, showing application potential in the phytoremediation of 4-Nitrobenzaldehyde-contaminated environments.
  • Widely used industrial chemical intermediate
  • Main photodegradation product of the antibiotic Chloramphenicol
  • Has genotoxic and mutagenic effects
  • Poses a threat to human health and ecosystems
  • Can be converted to a less toxic compound by specific transgenic rice
  • Potential for use in phytoremediation of contaminated environments