Learn More
Medchemexpress LLC 4-Nitrobenzaldehyde | 555-16-8 | 151.12 | 25 G
SDP
Catalog No. 5000449866
Encompass_Preferred
4-Nitrobenzaldehyde is a widely used industrial chemical intermediate and also the main photodegradation product of the broad-spectrum antibiotic Chloramphenicol. It has genotoxic and mutagenic effects and poses a certain threat to human health and ecosystems.
  • Widely used industrial chemical intermediate
  • Main photodegradation product of Chloramphenicol
  • Exhibits genotoxic and mutagenic effects
  • Can be converted to 4-aminobenzaldehyde with lower toxicity by EINFS1 transgenic rice for potential phytoremediation applications
Catalog No. 50-004-49866 Supplier Medchemexpress LLC Supplier No. 25GHYW007577
4-Nitrobenzaldehyde is a widely used industrial chemical intermediate and also the main photodegradation product of the broad-spectrum antibiotic Chloramphenicol. It has genotoxic and mutagenic effects and poses a certain threat to human health and ecosystems.
  • Widely used industrial chemical intermediate
  • Main photodegradation product of Chloramphenicol
  • Exhibits genotoxic and mutagenic effects
  • Can be converted to 4-aminobenzaldehyde with lower toxicity by EINFS1 transgenic rice for potential phytoremediation applications