Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzamidine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. N09895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09895G 5 g
N098925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09895G Supplier TCI America Supplier No. N09895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15723-90-7
Molecular Formula C7H8ClN3O2
Molecular Weight (g/mol) 201.61
MDL Number MFCD00278833
InChI Key CJXJAUKCHHAJQN-UHFFFAOYSA-N
PubChem CID 170625
IUPAC Name hydrogen 4-nitrobenzene-1-carboximidamide chloride
SMILES [H+].[Cl-].NC(=N)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 286°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzamidine Hydrochloride
RTECSNumber CV6296000
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.